missing translation for 'onlineSavingsMsg'
Learn More

Melford Ionomycin

Product Code. 30018354
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018354 5 mg 5mg
30018353 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30018354 Supplier Melford Supplier No. I520200.005
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ionomycin free acid is frequently employed in cell biology experiments to study cellular processes that are dependent on calcium signaling.

Ionomycin free acid is frequently employed in cell biology experiments to study cellular processes that are dependent on calcium signaling. By transporting calcium ions into cells, ionomycin can trigger various cellular responses, including the activation of enzymes and signaling pathways. Researchers use ionomycin in a variety of applications, such as studying cell signaling, apoptosis, and calcium-dependent processes. It is particularly useful in experiments where precise control over intracellular calcium levels is necessary.

Specifications

Chemical Name or Material Ionomycin
CAS 56092-81-0
Quantity 5 mg
Molecular Formula C41H72O9
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.