missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphate Standard for Ion Chromatography, SPEX CertiPrep™
Click to view available options
Quantity:
125 mL
500 mL
Packaging:
Triple-leached LDPE bottle
Description
Includes
NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
PO&subscript_4;&superscript_minus;&superscript_3; | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Packaging | Triple-leached LDPE bottle |
CAS Min % | 0.1, 99.9 |
Quantity | 500 mL |
CAS Max % | 0.1, 99.9 |
Certificate of Analysis | With Certificate of Analysis. |
Concentration or Composition Notes | Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. |
For Use With (Equipment) | AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation |
Concentration or Composition (by Analyte or Components) | 1000μg/mL |
Solvent or Matrix | H2O |
Chemical Name or Material | Phosphate Standard |
Safety and Handling
Product Identifier
- Volatile Organics Mix (High Level)
- Danger
- Acute toxicity Category 3
- Carcinogenicity Category 1A
- Flammable liquid Category 2
- Germ cell mutagenicity Category 1B
- Reproductive toxicity Category 1
- Skin sensitiser Category 1
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity after single exposure Category 1
- H225-Highly flammable liquid and vapour.
- H331-Toxic if inhaled.
- H340-May cause genetic defects.
- H350-May cause cancer.
- H360-May damage fertility or the unborn child.
- H370-Causes damage to organs.
- H373-May cause damage to organs.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : isopropyl benzene, methanol
Product intended for laboratory use only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'