missing translation for 'onlineSavingsMsg'
Learn More

Instrument Check Standard 3 For Ion Chromatography, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Brand:  SPEX Certiprep ICMIX3-100

Code : 04

Additional Details : CAS Number : 7697-37-2 Weight : 0.12500kg Transport : UN number : 3264 Chem class : 8 Pack group : III

Product Code. 11430209

Quantity Price
1 £156.00 / 125mL
6 £140.43 / 125mL
Estimated Shipment: 07-10-2021

Please to purchase this item. Need a web account? Register with us today!



Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Ca+21000μg/mL
NH4+400μg/mL
K+200μg/mL
Mg+2200μg/mL
Na+200μg/mL
Li+50μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Instrument Check Standard 3
Components in 2% HNO3:
Ca+2    •1000μg/mL
K+    •200μg/mL
Li+    •50μg/mL
Mg+2    •200μg/mL
Na+    •200μg/mL
NH4+    •400μg/mL
Triple-leached LDPE bottle
2% HNO3
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
With Certificate of Analysis
125mL
Videos
SDS
Documents
Certificates

Product intended for laboratory use only