missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ion Chromatography Fluoride Standard, 100μg/mL F-, SPEX CertiPrep™
Click to view available options
Quantity:
125 mL
500 mL
Packaging:
Poly Bottle
Triple-leached LDPE bottle
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
- Fluoride Standard
- Single-Element Ion Anion
- 100 μg/mL
- 500 mL volume
Certifications
ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015
Specifications
Packaging | Poly Bottle |
CAS Min % | 0.01, 99.99 |
Quantity | 500 mL |
CAS Max % | 0.01, 99.99 |
CAS | 7732-18-5 |
Certificate of Analysis | With Certificate of Analysis. |
Concentration or Composition (by Analyte or Components) | 100μg/mL F− |
Solvent or Matrix | H2O |
Chemical Name or Material | Ion Chromatography Fluoride Standard |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'