missing translation for 'onlineSavingsMsg'
Learn More

Simple Cyanide Reference Standard For Ion Chromatography, H2O/2% KOH, 1000μg/mL:KCN, SPEX CertiPrep™

Catalog Number 10192342
Click to view available options
Quantity:
125 mL
500 mL
Packaging:
Triple-leached LDPE bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
CN-1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

CAS Min % 0.1, 2.0, 97.9
CAS Max % 0.1, 2.0, 97.9
CAS 7732-18-5
Concentration or Composition (by Analyte or Components) For Use In    Matrix    Contents    Cat. No.
EPA Methods 335.2 and 335.3; ASTM Method D2036-19; and Standard Method 4500-CNF.    CN in H2O    1000μg/mL:KCN    RSCN9-2X, -2Y
EPA Method 335.1    CN in H2O    1000μg/mL:K3Fe(CN)6    RSCN9C-2X
Packaging Triple-leached LDPE bottle
Certificate of Analysis With Certificate of Analysis.
Solvent or Matrix 2% KOH
Quantity 125 mL
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Chemical Name or Material Cyanide Standard (Simple)
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1002
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H312-Harmful in contact with skin.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P405-Store locked up.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : potassium hydroxide,potassium cyanide

Product intended for laboratory use only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'