missing translation for 'onlineSavingsMsg'
Learn More

Ion 510™ Food Protection Chip Kit

Product Code. 16615595 Shop All Thermo Scientific Products
This item is not returnable. View return policy
Product Code. 16615595 Supplier Thermo Scientific Supplier No. A27769
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Simplify sample tracking and sequencing using this Chip Kit as part of a complete Thermo Scientific™ NGS Workflow for Food Authenticity.

Simplify sample tracking and sequencing with the Ion 510™ Food Protection Chip Kit as a part of  Thermo Scientific™ Next Generation Sequencing (NGS) Food Authenticity Workflow incorporating Ion Torrent™ Next Generation Sequencing technology.

The Thermo Scientific NGS Food Authenticity Workflow allows identification of meat, plant or fish species present in even the most challenging sample types, protecting your business from food fraud.

The Food Protection Ion 510 Chip Kit contains eight barcoded chips for sample tracking and sequencing with the Ion GeneStudio S5 Food Protection sequencing system.

Identification of the species present in food and feed samples is a critical step for proof of authenticity, verification of origin, traceability of raw materials, and quality control of handling and cleaning processes in food production lines. The Ion 510 Food Protection Chip Kit allows identification of meat, fish or plant species present in all food types as a part of complete NGS Food Authenticity Workflow.

  • Rapid: run times of only 2.5–4 hours, generating 2–3 million reads.
  • Simple: easy handling and loading of templated products for sequencing using the Ion Chef
  • Flexible: simultaneous analysis of multiple samples on a single chip using sample labeling with unique barcodes

The NGS Food Authenticity Workflow utilizes  Ion Torrent™ Next Generation Sequencing technology to enable an untargeted screening approach making it possible to identify the species contained in a food sample by comparison with a DNA database of meat, plant or fish species.

As a part of the Ion Torrent™ sequencing workflow, the Ion 510 Food Protection Chip Kit electronically detects polymerase-driven base incorporation without the use of fluorescence and offers easy handling and loading of templated products for sequencing.

For Research Use Only. Not for use in diagnostic procedures.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Room Temperature
Description Ion 510 Food Protection Chip Kit
Format Kit
For Use With (Application) Sequencing
Packaging Type 2 boxes
Product Type Food Protection Chip Kit
Quantity 8 chips
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.