missing translation for 'onlineSavingsMsg'
Learn More

Iohexol, Mikromol™

Product Code. 17989464
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17989464 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17989464 Supplier Mikromol Supplier No. MM1468.00
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Iohexol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Iohexol
CAS 66108-95-0
Analyte or Component Names Iohexol
Shipping Condition Room Temperature
Molecular Formula C19 H26 I3 N3 O9
Certifications/Compliance ISO 17025
Synonym Iohexol, 5-[Acetyl(2,3-dihydroxypropyl)amino]-N,N'-bis(2,3-dihydroxypropyl)-2,4,6-triiodobenzene-1,3-dicarboxamide, 1,3-Benzenedicarboxamide, 5-[acetyl(2,3-dihydroxypropyl)amino]-N,N'-bis(2,3-dihydroxypropyl)-2,4,6-triiodo- (9CI), 5-(N-2,3-Dihydroxypropylacetamido)-2,4,6-triiodo-N,N'-bis(2,3-dihydroxypropyl)isophthalamide., Accudenz, Accupaque 300, Exypaque, Histodenz, Iopaque 240, Iopaque 300, Ioverin 350, Nycodenz, Omnipaque, Omnipaque 140, Omnipaque 240, Omnipaque 300, Omnipaque 350, Omnipaque R
InChI Formula InChI=1S/C19H26I3N3O9/c1-8(29)25(4-11(32)7-28)17-15(21)12(18(33)23-2-9(30)5-26)14(20)13(16(17)22)19(34)24-3-10(31)6-27/h9-11,26-28,30-32H,2-7H2,1H3,(H,23,33)(H,24,34)
SMILES CC(=O)N(CC(O)CO)c1c(I)c(C(=O)NCC(O)CO)c(I)c(C(=O)NCC(O)CO)c1I
IUPAC Name 5-[acetyl(2,3-dihydroxypropyl)amino]-1-N,3-N-bis(2,3-dihydroxypropyl)-2,4,6-triiodobenzene-1,3-dicarboxamide
Molecular Weight (g/mol) 821.14
Quantity 250 mg
Formula Weight 820.8803 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.