missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iohexol, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
Iohexol
Specifications
Specifications
| Chemical Name or Material | Iohexol |
| CAS | 66108-95-0 |
| Analyte or Component Names | Iohexol |
| Shipping Condition | Room Temperature |
| Molecular Formula | C19 H26 I3 N3 O9 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Iohexol, 5-[Acetyl(2,3-dihydroxypropyl)amino]-N,N'-bis(2,3-dihydroxypropyl)-2,4,6-triiodobenzene-1,3-dicarboxamide, 1,3-Benzenedicarboxamide, 5-[acetyl(2,3-dihydroxypropyl)amino]-N,N'-bis(2,3-dihydroxypropyl)-2,4,6-triiodo- (9 CI), 5-(N-2,3-Dihydroxypropylacetamido)-2,4,6-triiodo-N,N'-bis(2,3-dihydroxypropyl)isophthalamide., Accudenz, Accupaque 300, Exypaque, Histodenz, Iopaque 240, Iopaque 300, Ioverin 350, Nycodenz, Omnipaque, Omnipaque 140, Omnipaque 240, Omnipaque 300, Omnipaque 350, Omnipaque R |
| InChI Formula | InChI=1 S/C19H26I3N3O9/c1-8(29)25(4-11(32)7-28)17-15(21)12(18(33)23-2-9(30)5-26)14(20)13(16(17)22)19(34)24-3-10(31)6-27/h9-11,26-28,30-32 H,2-7H2,1H3,(H,23,33)(H,24,34) |
| SMILES | CC(=O)N(CC(O)CO)c1c(I)c(C(=O)NCC(O)CO)c(I)c(C(=O)NCC(O)CO)c1I |
| IUPAC Name | 5-[acetyl(2,3-dihydroxypropyl)amino]-1-N,3-N-bis(2,3-dihydroxypropyl)-2,4,6-triiodobenzene-1,3-dicarboxamide |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction