missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| Packaging | Glass Bottle |
| Analyte or Component Names | Iodoplatinate Reagent. |
| Purity Grade Notes | EU Ph Ref: 1046300 |
| Element | Iodoplatinate |
| Quantity | 200 mL |
| Grade | Eu. Ph. |
Safety and Handling
Product Identifier
- Iodoplatinate Reagent. European Pharmacopoeia Ref: 1046300 (RP 108)
- Danger
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
- Reproductive toxicity Category 2
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H361-Suspected of damaging fertility or the unborn child.
- P202-Do not handle until all safety precautions have been read and understood.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P501-Dispose of contents/container to .
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
