missing translation for 'onlineSavingsMsg'
Learn More

Iodoplatinate Reagent According to European Pharmacopoeia (EP), Reagecon
BIOPHARMA

Product Code. 11404699 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
200 mL
Unit Size:
200mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11404699 200 mL 200mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11404699 Supplier Reagecon Supplier No. 1046300

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity, stable product developed and tested for European Pharmacopoeia (EP) applications including use as a reagent in the performance of tests outlined in several EP monographs.

  • European Pharmacopoeia Reagent, Solution
  • High-purity, stable product developed and tested for European Pharmacopoeia (EP) applications.
  • Applications include use as a reagent in the performance of tests outlined in several EP monographs.
  • Accurate, stable and reproducible.
  • Stability is proven (either opened or unopened) for the shelf life of this product.

Specifications

Packaging Amber Glass Winchester Bottle
Physical Form Liquid
Technique EP Testing
Quantity 200 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.