missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodopanoic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | Iopanoic acid |
| CAS | 96-83-3 |
| Molecular Formula | C11 H12 I3 N O2 |
| Quantity | 1 g |
| Synonym | Iopanoic acid, (RS)-2-(3-amino-2,4,6-tri-iodobenzyl)butanoic acid, 2-(3-Amino-2,4,6-triiodobenzyl)butyric acid, 2-Ethyl-3-(3-amino-2,4,6-triiodophenyl)propionic acid, 2-[(5-Amino-2,4,6-triiodophenyl)methyl]butanoic acid, 3-(3-Amino-2,4,6-triiodophenyl)-2-ethylpropanoic acid, 3-(3-Amino-2,4,6-triiodophenyl)-2-ethylpropionic acid, Bilijodon, Choladine, Cholevid, Cistobil, Colepax, Copanoic, Iodopanic acid, Iodopanoic acid, Iopagnost, Iopanoicum, Jopagnost, NSC 41706, Telepaque, 3-amino-alpha-ethyl-2,4,6-triiodobenzenepropanoic acid, Hydrocinnamic acid, 3-amino-alpha-ethyl-2,4,6-triiodo- (6CI,7CI,8CI), (+/-)-Iopanoic acid, 3-Amino-alpha-ethyl-2,4,6-triiodohydrocinnamic acid, alpha-Ethyl-beta-(3-amino-2,4,6-triiodophenyl)propionic acid |
| InChI Formula | InChI=1S/C11H12I3NO2/c1-2-5(11(16)17)3-6-7(12)4-8(13)10(15)9(6)14/h4-5H,2-3,15H2,1H3,(H,16,17) |
| SMILES | CCC(Cc1c(I)cc(I)c(N)c1I)C(=O)O |
| IUPAC Name | 2-[(3-amino-2,4,6-triiodophenyl)methyl]butanoic acid |
| Molecular Weight (g/mol) | 570.93 |
| Formula Weight | 570.8002 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?