missing translation for 'onlineSavingsMsg'
Learn More

Iodopanoic Acid, TRC

Product Code. 30395686
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30395686 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30395686 Supplier Toronto Research Chemicals Supplier No. TRCI71740010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Iopanoic acid
CAS 96-83-3
Molecular Formula C11 H12 I3 N O2
Quantity 10 g
Synonym Iopanoic acid, (RS)-2-(3-amino-2,4,6-tri-iodobenzyl)butanoic acid, 2-(3-Amino-2,4,6-triiodobenzyl)butyric acid, 2-Ethyl-3-(3-amino-2,4,6-triiodophenyl)propionic acid, 2-[(5-Amino-2,4,6-triiodophenyl)methyl]butanoic acid, 3-(3-Amino-2,4,6-triiodophenyl)-2-ethylpropanoic acid, 3-(3-Amino-2,4,6-triiodophenyl)-2-ethylpropionic acid, Bilijodon, Choladine, Cholevid, Cistobil, Colepax, Copanoic, Iodopanic acid, Iodopanoic acid, Iopagnost, Iopanoicum, Jopagnost, NSC 41706, Telepaque, 3-amino-alpha-ethyl-2,4,6-triiodobenzenepropanoic acid, Hydrocinnamic acid, 3-amino-alpha-ethyl-2,4,6-triiodo- (6CI,7CI,8CI), (+/-)-Iopanoic acid, 3-Amino-alpha-ethyl-2,4,6-triiodohydrocinnamic acid, alpha-Ethyl-beta-(3-amino-2,4,6-triiodophenyl)propionic acid
InChI Formula InChI=1S/C11H12I3NO2/c1-2-5(11(16)17)3-6-7(12)4-8(13)10(15)9(6)14/h4-5H,2-3,15H2,1H3,(H,16,17)
SMILES CCC(Cc1c(I)cc(I)c(N)c1I)C(=O)O
IUPAC Name 2-[(3-amino-2,4,6-triiodophenyl)methyl]butanoic acid
Molecular Weight (g/mol) 570.93
Formula Weight 570.8002
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.