missing translation for 'onlineSavingsMsg'
Learn More

(Iodomethyl)trimethylsilane, 99%

Product Code. 11461960
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11461960 50 g 50g
11451960 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11461960 Supplier Thermo Scientific Alfa Aesar Supplier No. A15120.18
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4206-67-1 | C4H11ISi | 214.121 g/mol

(Iodomethyl)trimethylsilane is used for the N-alkylation of amides, in a reaction sequence to prepare unstabilized ylides for [2+3]-cycloadditions. It can react with acetoacetic acid ethyl ester to get 3-trimethylsilanyl-proπonic acid ethyl ester.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(Iodomethyl)trimethylsilane is used for the N-alkylation of amides, in a reaction sequence to prepare unstabilized ylides for [2+3]-cycloadditions. It can react with acetoacetic acid ethyl ester to get 3-trimethylsilanyl-proπonic acid ethyl ester.

Solubility
Difficult to mix.

Notes
Light Sensitive. Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4206-67-1
Molecular Formula C4H11ISi
Molecular Weight (g/mol) 214.121
MDL Number MFCD00001077
InChI Key VZNYXGQMDSRJAL-UHFFFAOYSA-N
Synonym iodomethyl trimethylsilane, iodomethyl-trimethylsilane, iodomethyl trimethyl silane, trimethylsilyl methyl iodide, sila-neopentyliodide, trimethylsilylmethyl iodide, acmc-209jn7, trimethyl iodomethyl silane, iodanylmethyl trimethyl silane
PubChem CID 77877
IUPAC Name iodomethyl(trimethyl)silane
SMILES C[Si](C)(C)CI

Specifications

Boiling Point 140°C to 142°C
Quantity 50 g
Linear Formula (CH3)3SiCH2I
UN Number UN1993
Beilstein 1731485
Flash Point 31°C (88°F)
Solubility Information Difficult to mix.
Formula Weight 214.12
Percent Purity 99%
Odor Recognizable
Refractive Index 1.4915
Sensitivity Light sensitive
Density 1.443
Chemical Name or Material (Iodomethyl)trimethylsilane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.