missing translation for 'onlineSavingsMsg'
Learn More

Iodomethyl Pivalate, TRC

Product Code. 30399680
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30399680 1 g
30406846 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30399680 Supplier Toronto Research Chemicals Supplier No. TRCI7087501G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53064-79-2
Molecular Formula C6 H11 I O2
Molecular Weight (g/mol) 242.05
Synonym 2,2-Dimethyl-propanoic Acid Iodomethyl Ester, (Pivaloyloxy)methyl Iodide, 2,2-Dimethylpropionic Acid Iodomethyl Ester, Iodomethyl 2,2-Dimethylpropanoate
IUPAC Name iodomethyl 2,2-dimethylpropanoate
SMILES CC(C)(C)C(=O)OCI

Specifications

Quantity 1 g
InChI Formula InChI=1S/C6H11IO2/c1-6(2,3)5(8)9-4-7/h4H2,1-3H3
Formula Weight 241.9804
Chemical Name or Material Iodomethyl pivalate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.