missing translation for 'onlineSavingsMsg'
Learn More

(Iodomethyl)cyclopropane, 95%

Product Code. p-7032892
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15497497

Brand: Thermo Scientific Alfa Aesar H35309.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 33574-02-6 | C4H7I | 182.004 g/mol

(Iodomethyl)cyclopropane is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(Iodomethyl)cyclopropane is used as pharmaceutical intermediate.

Notes
Light sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33574-02-6
Molecular Formula C4H7I
Molecular Weight (g/mol) 182.004
MDL Number MFCD06410734
InChI Key JHKJQWFHNIOUKY-UHFFFAOYSA-N
Synonym iodomethyl cyclopropane, cyclopropylmethyl iodide, cyclopropane, iodomethyl, ambkkkkk549, acmc-20aplj, 2-iodomethyl cyclopropane, iodomethyl cyclopropane gc
PubChem CID 2763048
IUPAC Name iodomethylcyclopropane
SMILES C1CC1CI

Specifications

Quantity 250 mg
Sensitivity Light sensitive
Formula Weight 182
Percent Purity 97%
Chemical Name or Material (Iodomethyl)cyclopropane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.