missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodomethane, 99%, stabilized, Thermo Scientific Chemicals
76.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
72.38 GBP valid until 2024-07-31
Use promo code "23251" to get your promotional price.
Use promo code "22914" to get your promotional price.
72.38 GBP valid until 2024-07-31
Use promo code "23251" to get your promotional price.
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagent
Specifications
Melting Point | -66.0°C |
Density | 2.2800g/mL |
Boiling Point | 42.5°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% |
Packaging | Glass bottle |
Linear Formula | CH3I |
Refractive Index | 1.5290 to 1.5310 |
Quantity | 500 g |
Beilstein | 01, 69 |
Show More |
Safety and Handling
Product Identifier
- Iodomethane
- Danger
- Acute toxicity Category 3
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Carcinogenicity Category 2
- Specific target organ toxicity Category 3
- H301-Toxic if swallowed.
- H312-Harmful in contact with skin.
- H315-Causes skin irritation.
- H331-Toxic if inhaled.
- H335-May cause respiratory irritation.
- H351-Suspected of causing cancer.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P311-Call a POISON CENTER/doctor/ .
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'