missing translation for 'onlineSavingsMsg'
Learn More

Iodomethane-d3, for NMR, 99+ atom % D

Product Code. 10790871 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10790871 25 g Glass Bottle 25g
10780871 5 g Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10790871 Supplier Thermo Scientific Acros Supplier No. 174950250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 865-50-9 | CH3I | 144.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 865-50-9
Molecular Formula CH3I
Molecular Weight (g/mol) 144.96
MDL Number MFCD00001074
InChI Key INQOMBQAUSQDDS-FIBGUPNXSA-N
Synonym iodomethane-d3, iodo 2h3 methane, trideuterio iodo methane, iodo 2 h? methane, methyl-d3 iodide, trideuteromethyl iodide, iodomethane-12c,d3, methyl-12c,d3 iodide, iodoethane-d5 ethyliodid-d5, methyl iodide d3 stored over copper
PubChem CID 2723978
IUPAC Name trideuterio(iodo)methane
SMILES [2H]C([2H])([2H])I

Specifications

Physical Form Liquid
Color Colorless
Density 2.3290g/mL
Deuterium (D) 99.0% min.
Packaging Glass Bottle
Refractive Index 1.5262
Water 0.05% max. (Coulometric)
Quantity 25 g
Boiling Point 42.0°C
Linear Formula CD3I
Formula Weight 144.96
Grade NMR
Chemical Name or Material Iodomethane-d3, 99+ atom % D
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Iodomethane-d3
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H330-Fatal if inhaled.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.