missing translation for 'onlineSavingsMsg'
Learn More

Iodomethane, 99%, stabilized

Product Code. p-4509695 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
100g
2.5kg
500g
5g
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

Product Code. 10677272

Brand: Thermo Scientific Acros 122370050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

CAS: 74-88-4 | CH3I | 141.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74-88-4, 7440-50-8
Molecular Formula CH3I
Molecular Weight (g/mol) 141.94
MDL Number MFCD00001073
InChI Key INQOMBQAUSQDDS-UHFFFAOYSA-N
Synonym methyl iodide, monoiodomethane, methane, iodo, methyljodid, jod-methan, methyliodide, methyljodide, iodometano, joodmethaan, metylu jodek
PubChem CID 6328
ChEBI CHEBI:39282
IUPAC Name iodomethane
SMILES CI

Specifications

Melting Point -66.0°C
Density 2.2800g/mL
Boiling Point 42.5°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3I
Refractive Index 1.5290 to 1.5310
Quantity 5 g
Beilstein 01, 69
Fieser 01,682; 02,274; 03,202; 04,341; 05,447; 06,384; 08,342; 09,308; 15,118
Merck Index 15, 6159
Specific Gravity 2.28
Solubility Information Solubility in water: 14g/L (20°C). Other solubilities: miscible with alcohol and ether, soluble in benzene, acetone, methylenechloride, n,n-dimethylformamide
Stabilizer Copper
Viscosity 0.5 mPa.s (20°C)
Formula Weight 141.94
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Iodomethane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Iodomethane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P311-Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contains : Iodomethane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.