missing translation for 'onlineSavingsMsg'
Learn More

Iodomethane, 99%, stab. with copper

Product Code. 11420980
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11420980 1000 g 1000g
11620222 50 g 50g
11440980 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11420980 Supplier Thermo Scientific Alfa Aesar Supplier No. A10559.0B
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 74-88-4 | CH3I | 141.94 g/mol

Iodomethane is a methylating agent used for alkylating carbon, oxygen, sulfur, nitrogen and phosphorus nucleophiles. For example, phenol reacts with methyl iodide to give anisole. In the Monsanto process, the insitu formed iodomethane reacts with carbon monoxide in the presence of a rhodium complex to give acetyl iodide, which on hydrolysis yields acetic acid. It reacts with magnesium to form the Grignard reagent, methylmagnesium iodide used in organic synthesis. It finds applications as an intermediate in the manufacture of pharmaceuticals and in phase transfer catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iodomethane is a methylating agent used for alkylating carbon, oxygen, sulfur, nitrogen and phosphorus nucleophiles. For example, phenol reacts with methyl iodide to give anisole. In the Monsanto process, the insitu formed iodomethane reacts with carbon monoxide in the presence of a rhodium complex to give acetyl iodide, which on hydrolysis yields acetic acid. It reacts with magnesium to form the Grignard reagent, methylmagnesium iodide used in organic synthesis. It finds applications as an intermediate in the manufacture of pharmaceuticals and in phase transfer catalysts.

Solubility
Miscible with water.

Notes
Light and moisture sensitive. Store in cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong bases and oxygen.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74-88-4
Molecular Formula CH3I
Molecular Weight (g/mol) 141.94
MDL Number MFCD00001073
InChI Key INQOMBQAUSQDDS-UHFFFAOYSA-N
Synonym methyl iodide, monoiodomethane, methane, iodo, methyljodid, jod-methan, methyliodide, methyljodide, iodometano, joodmethaan, metylu jodek
PubChem CID 6328
ChEBI CHEBI:39282
IUPAC Name iodomethane
SMILES CI

Specifications

Melting Point -67°C
Density 2.28
Boiling Point 41°C to 43°C
Odor Pungent
Refractive Index 1.53
Quantity 1000 g
UN Number UN2644
Beilstein 969135
Sensitivity Light sensitive
Merck Index 14,6087
Solubility Information Miscible with water.
Formula Weight 141.94
Percent Purity 99%
Chemical Name or Material Iodomethane, Stabilized with copper
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Iodomethane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P311-Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contain: Iodomethane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.