missing translation for 'onlineSavingsMsg'
Learn More

Iodomethane, 2M soln. in tert-butyl methyl ether

Product Code. 15458155
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15458155 500 mL 500mL
15448155 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15458155 Supplier Thermo Scientific Alfa Aesar Supplier No. H32660.AP

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 74-88-4 | CH3I | 141.94 g/mol

Iodomethane is used to control insects, plant parasitic nematodes, soil borne pathogens and weed seeds. Iodomethane as a potential metal mobilizing agent in nature. Microhydration effects on the intermediates of the S(N)2 reaction of iodide anion with methyl iodide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iodomethane is used to control insects, plant parasitic nematodes, soil borne pathogens and weed seeds. Iodomethane as a potential metal mobilizing agent in nature. Microhydration effects on the intermediates of the S(N)2 reaction of iodide anion with methyl iodide.

Solubility
Insoluble in water.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat, light, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74-88-4
Molecular Formula CH3I
Molecular Weight (g/mol) 141.94
MDL Number MFCD00001073
InChI Key INQOMBQAUSQDDS-UHFFFAOYSA-N
Synonym methyl iodide, monoiodomethane, methane, iodo, methyljodid, jod-methan, methyliodide, methyljodide, iodometano, joodmethaan, metylu jodek
PubChem CID 6328
ChEBI CHEBI:39282
IUPAC Name iodomethane
SMILES CI

Specifications

Density 0.933
Boiling Point 41°C to 43°C
Quantity 500 mL
UN Number UN1992
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 141.94
Concentration or Composition (by Analyte or Components) 2M soln. in tert-butyl methyl ether
Chemical Name or Material Iodomethane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.