missing translation for 'onlineSavingsMsg'
Learn More

Iodoform, 99+%

Product Code. 10413835 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10413835 2.5 kg Glass bottle 2.5kg
10543311 100 g Glass bottle 100g
10234720 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10413835 Supplier Thermo Scientific Acros Supplier No. 149120025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 75-47-8 | CHI3 | 393.72 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-47-8
Molecular Formula CHI3
Molecular Weight (g/mol) 393.72
MDL Number MFCD00001069
InChI Key OKJPEAGHQZHRQV-UHFFFAOYSA-N
Synonym triiodomethane, methane, triiodo, carbon triiodide, jodoform, trijodmethane, dezinfekt v, jodoform czech, trijodmethane czech, ccris 346, unii-kxi2j76489
PubChem CID 6374
ChEBI CHEBI:37758
IUPAC Name iodoform
SMILES C(I)(I)I

Specifications

Melting Point 119.0°C to 122.0°C
Color Yellow
Ignition Residue 0.1% max.
Loss on Drying 0.5% max. (24 hrs) (over silicagel)
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula CHI3
Quantity 2.5 kg
Beilstein 01, 73
Fieser 03,165; 04,266
Merck Index 15, 5076
Solubility Information Solubility in water: 0.1g/L (20°C). Other solubilities: 1g/60mL cold alcohol- 1g/16mL boiling alcohol,1g/10mL chloroform- 1g/7.5mL ether,1g/80mL glycerol- 1g/3mL cs2,freely soluble in benzene and acetone
Formula Weight 393.72
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Iodoform, 99+%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.