missing translation for 'onlineSavingsMsg'
Learn More

Iodoethane (Stabilized with Copper), TRC

Product Code. 17974165
Change view
Click to view available options
Quantity:
10 g
100 g
Unit Size:
100g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17974165 100 g 100g
17972825 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17974165 Supplier Toronto Research Chemicals Supplier No. TRCI705870100G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-03-6
Molecular Formula C2H5I
Molecular Weight (g/mol) 155.97
Synonym 1-Iodoethane, Ethyl Iodide, Hydriodic Ether, NSC 8825, Monoiodoethane
IUPAC Name iodoethane
SMILES CCI

Specifications

Quantity 100 g
InChI Formula InChI=1S/C2H5I/c1-2-3/h2H2,1H3
Formula Weight 155.9436
Chemical Name or Material Iodoethane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.