missing translation for 'onlineSavingsMsg'
Learn More

Iodocyclohexane, 98%, stab. with copper

Product Code. 11436626
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11436626 25 g 25g
11446626 100 g 100g
11456626 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11436626 Supplier Thermo Scientific Alfa Aesar Supplier No. B24840.14
  • ONLINE EXCLUSIVE PRICE
    £37.40
    £33.65 / 25g
    Save £3.75
    10% Off
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 626-62-0 | C6H11I | 210.06 g/mol

Iodocyclohexane has been employed as reagent in demethylation of aryl methyl ethers in DMF under reflux condition.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iodocyclohexane has been employed as reagent in demethylation of aryl methyl ethers in DMF under reflux condition.

Solubility
Soluble in ethanol, ether, acetone.

Notes
Light sensitive. Store away from oxidizing agent, light.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 626-62-0
Molecular Formula C6H11I
Molecular Weight (g/mol) 210.06
MDL Number MFCD00003826
InChI Key FUCOMWZKWIEKRK-UHFFFAOYSA-N
Synonym cyclohexane, iodo, cyclohexyl iodide, cyclohexyliodide, iodo-cyclohexane, 1-iodocyclohexane, heptynyltrimethylsilane, iodocyclohexane, acmc-1att4, c-c6h11i
PubChem CID 12289
IUPAC Name iodocyclohexane
SMILES IC1CCCCC1

Specifications

Density 1.625
Boiling Point 180°C
Flash Point 71°C (159°F)
Refractive Index 1.5475
Quantity 25 g
Beilstein 1900797
Sensitivity Light sensitive
Solubility Information Soluble in ethanol,ether,acetone.
Formula Weight 210.06
Percent Purity 98%
Chemical Name or Material Iodocyclohexane, stabilized with copper
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.