missing translation for 'onlineSavingsMsg'
Learn More

Iodoacetyl chloride, 97%

Product Code. 11361577
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11361577

Brand: Thermo Scientific Alfa Aesar L04428.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 38020-81-4 | C2H2ClIO | 204.39 g/mol

Iodoacetyl chloride is used for biochemical research purposes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iodoacetyl chloride is used for biochemical research purposes.

Solubility
Miscible with water.

Notes
Moisture and light sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong bases and strong reducing agents. Reacts with water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38020-81-4
Molecular Formula C2H2ClIO
Molecular Weight (g/mol) 204.39
MDL Number MFCD00013718
InChI Key BSVMPWANOMFSPR-UHFFFAOYSA-N
Synonym iodoacetyl chloride, iodoacetylchloride, acetyl chloride, iodo, jodacetylchlorid, acetylchloride,iodo, iodoacetic acid chloride, acmc-1aewu, iodoacetyl chloride 5g, acetyl chloride, iodo-6ci,9ci
PubChem CID 3084680
IUPAC Name 2-iodoacetyl chloride
SMILES ClC(=O)CI

Specifications

Density 2.28
Boiling Point 48°C to 50°C (12 mmHg)
Quantity 25 g
UN Number UN3265
Beilstein 1739078
Sensitivity Light and moisture sensitive
Solubility Information Miscible with water.
Formula Weight 204.39
Percent Purity 97%
Chemical Name or Material Iodoacetyl chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.