missing translation for 'onlineSavingsMsg'
Learn More

Iodoacetyl chloride, 97%

Product Code. 11351577
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11351577 5 g 5g
11361577 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11351577 Supplier Thermo Scientific Alfa Aesar Supplier No. L04428.06
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 38020-81-4 | C2H2ClIO | 204.39 g/mol

Iodoacetyl chloride is used for biochemical research purposes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iodoacetyl chloride is used for biochemical research purposes.

Solubility
Miscible with water.

Notes
Moisture and light sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong bases and strong reducing agents. Reacts with water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38020-81-4
Molecular Formula C2H2ClIO
Molecular Weight (g/mol) 204.39
MDL Number MFCD00013718
InChI Key BSVMPWANOMFSPR-UHFFFAOYSA-N
Synonym iodoacetyl chloride, iodoacetylchloride, acetyl chloride, iodo, jodacetylchlorid, acetylchloride,iodo, iodoacetic acid chloride, acmc-1aewu, iodoacetyl chloride 5g, acetyl chloride, iodo-6ci,9ci
PubChem CID 3084680
IUPAC Name 2-iodoacetyl chloride
SMILES ClC(=O)CI

Specifications

Density 2.28
Boiling Point 48°C to 50°C (12 mmHg)
Quantity 5 g
UN Number UN3265
Beilstein 1739078
Sensitivity Light and moisture sensitive
Solubility Information Miscible with water.
Formula Weight 204.39
Percent Purity 97%
Chemical Name or Material Iodoacetyl chloride
compliance-icons
Product Identifier
  • Iodoacetyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Iodoacetyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.