missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Vivaspin™ 500 Centrifugal Concentrators
BIOPHARMA

Product Code. 10499701 Shop All Sartorius Products
Change view
Click to view available options
MWCO:
0.2 μm
10 kDa
100 kDa
1000 kDa
3 kDa
30 kDa
300 kDa
5 kDa
5, 10, 30, 50, and 100 kDa
50 kDa
Quantity:
100/Pk.
25/Pk.
Unit Size:
Pack of 100
Pack of 25
19 product options available for selection
Product selection table with 19 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. MWCO Quantity unitSize
10499701 300 kDa 25/Pk. Pack of 25
10019460 10 kDa 25/Pk. Pack of 25
10719001 5 kDa 100/Pk. Pack of 100
10029510 5 kDa 25/Pk. Pack of 25
10125580 10 kDa 100/Pk. Pack of 100
10681842 30 kDa 25/Pk. Pack of 25
10029470 50 kDa 25/Pk. Pack of 25
10375361 100 kDa 25/Pk. Pack of 25
10459512 50 kDa 100/Pk. Pack of 100
10073930 30 kDa 100/Pk. Pack of 100
10223621 100 kDa 100/Pk. Pack of 100
10767052 300 kDa 100/Pk. Pack of 100
10727632 1000 kDa 25/Pk. Pack of 25
10174281 1000 kDa 100/Pk. Pack of 100
10184281 0.2 μm 25/Pk. Pack of 25
10658961 0.2 μm 100/Pk. Pack of 100
10440282 3 kDa 25/Pk. Pack of 25
10488742 5, 10, 30, 50, and 100 kDa 25/Pk. Pack of 25
10689151 3 kDa 100/Pk. Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 19 options available.
19 options
This item is not returnable. View return policy
Product Code. 10499701 Supplier Sartorius Supplier No. VS0151
  • ONLINE EXCLUSIVE PRICE
    £125.33
    £109.00 / Pack of 25
    Save £16.33
    13% Off
View all promotions
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Provides a simple, one-step concentration procedure. Sartorius Vivaspin™ 500 Centrifugal Concentrators are disposable ultrafiltration devices for the concentration of biological samples with starting volumes up to 500μL. Available with PES membranes in a wide range of molecular weight cut-offs.

  • Patented vertical membrane design and thin channel filtration chamber minimizes membrane fouling and provides high speed concentrations
  • Concentrate recoveries of > 90% can be reached in less than 30 minutes
  • The dead stop pocket prevents concentration to dryness for safe sample concentration
  • Universal rotor compatibility: can be used in fixed angle rotors accepting 2.2mL centrifuge tubes
  • High-flux PES membranes for ultra-fast concentration
  • Convenient concentrate recovery by pipetting
  • Clear housing with volume graduation for easy reading/monitoring
TRUSTED_SUSTAINABILITY

Specifications

Disposable Single-use
Filter Type Ultrafiltration
For Use With (Equipment) Centrifuge
Material (Membrane) PES
MWCO 300 kDa
Sample Volume 500 mL
Sterility Non-sterile
Filtration Area 64.5m m2
Housing Material Polycarbonate
Quantity 25/Pk.
Volume (Metric) 0.5 mL
Length (Metric) 50 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.