missing translation for 'onlineSavingsMsg'
Learn More

Iodixanol, TRC

Product Code. 30133637
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133637 100 mg 100mg
30006732 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133637 Supplier Toronto Research Chemicals Supplier No. TRCI667200100MG
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92339-11-2
Molecular Formula C35 H44 I6 N6 O15
Molecular Weight (g/mol) 1550.18
Synonym 5,5'-[(2-Hydroxy-1,3-propanediyl)bis(acetylimino)]bis[N,N'-bis(2,3-dihydroxypropyl)-2,4,6-triiodo-1,3-benzenedicarboxamide, 2-5410-3A, Iodixanol, OptiPrep, Visipaque, Visipaque 270
IUPAC Name 5-[acetyl-[3-[N-acetyl-3,5-bis(2,3-dihydroxypropylcarbamoyl)-2,4,6-triiodoanilino]-2-hydroxypropyl]amino]-1-N,3-N-bis(2,3-dihydroxypropyl)-2,4,6-triiodobenzene-1,3-dicarboxamide
SMILES CC(=O)N(CC(O)CN(C(=O)C)c1c(I)c(C(=O)NCC(O)CO)c(I)c(C(=O)NCC(O)CO)c1I)c2c(I)c(C(=O)NCC(O)CO)c(I)c(C(=O)NCC(O)CO)c2I

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C35H44I6N6O15/c1-13(52)46(30-26(38)20(32(59)42-3-15(54)9-48)24(36)21(27(30)39)33(60)43-4-16(55)10-49)7-19(58)8-47(14(2)53)31-28(40)22(34(61)44-5-17(56)11-50)25(37)23(29(31)41)35(62)45-6-18(57)12-51/h15-19,48-51,54-58H,3-12H2,1-2H3,(H,42,59)(H,43,60)(H,44,61)(H,45,62)
Formula Weight 1549.7133
Chemical Name or Material Iodixanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.