missing translation for 'onlineSavingsMsg'
Learn More

Iodipamide, MedChemExpress

Product Code. 16488688
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16488688 10 mg 10mg
16454577 50 mg 50mg
16411108 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16488688 Supplier Medchem Express Supplier No. HYB1292
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Iodipamide is a tri-iodinated benzoate derivative and ionic dimeric contrast agent used in diagnostic imaging.

Iodipamide is a tri-iodinated benzoate derivative and ionic dimeric contrast agent used in diagnostic imaging.

Specifications

Chemical Name or Material Iodipamide
CAS 606-17-7
Color Light Nude
Purity Grade Notes Research
Molecular Formula C20H14I6N2O6
Quantity 10 mg
Synonym Adipiodone
Solubility Information DMSO : 125 mg/mL (109.67 mM; Need ultrasonic)
SMILES O=C(NC1=C(I)C(C(O)=O)=C(I)C=C1I)CCCCC(NC2=C(I)C(C(O)=O)=C(I)C=C2I)=O
Molecular Weight (g/mol) 1139.76
Formula Weight 1139.76
Percent Purity 99.08%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.