missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodine trichloride, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Iodine trichloride can be used to introduce iodine and chlorine in organic synthesis. It is also used as topical antiseptic and as laboratory reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Iodine trichloride can be used to introduce iodine and chlorine in organic synthesis. It is also used as topical antiseptic and as laboratory reagent.
Solubility
Soluble with decomposition.
Notes
Light Sensitive. Store at -20°C.
Iodine trichloride can be used to introduce iodine and chlorine in organic synthesis. It is also used as topical antiseptic and as laboratory reagent.
Solubility
Soluble with decomposition.
Notes
Light Sensitive. Store at -20°C.
Specifications
Quantity | 100 g |
Linear Formula | ICl3 |
UN Number | UN3085 |
Merck Index | 14,5020 |
Solubility Information | Soluble with decomposition. |
Formula Weight | 233.26 |
Percent Purity | 97% |
Sensitivity | Light sensitive |
Chemical Name or Material | Iodine trichloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'