missing translation for 'onlineSavingsMsg'
Learn More

Iodine spheres, ultra dry, -10 mesh, 99.998% (metals basis)

Product Code. 11377877
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11377877

Brand: Thermo Scientific Alfa Aesar 014312.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7553-56-2 | I{2}

Iodine spheres are used in OLED materials, chemicals, pharmaceuticals and electronics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 183°C to 186°C (sublimation)
CAS 7553-56-2
Physical Form Solid
Quantity 25 g
Molecular Formula I{2}
Percent Purity 99.998%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.