missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodine Solution 1% (w/v) In Potassium Iodide, Pure, Fisher Chemical™
Shop All Fisher Chemical ProductsDescription
Iodine Solution 1% (w/v) In Potassium Iodide, Pure
Specifications
Specifications
| Color | Amber |
| Packaging | Amber glass bottle |
| Quantity | 1 L |
| Physical Form | Liquid |
Safety and Handling
Product Identifier
- Iodine solution 1% w/v in potassium iodide
- Warning
- Specific target organ toxicity after repeated exposure Category 2
- H373-May cause damage to organs.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P314-Get medical advice/attention if you feel unwell.
- MIXTURE LIST-Contain: Iodine
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?