missing translation for 'onlineSavingsMsg'
Learn More

Iodine Solution 0.05M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™

Product Code. p-7073146 Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 L
2.5 L
Unit Size:
1 L
2.5 L
This item is not returnable. View return policy

Product Code. 10625112

Brand: Fisher Chemical J/4410/15

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

I2, CAS Number-7553-56-2, tincture iodine, iodine sublimed, iodine solution, iodine crystals, iodine, iode, diiodine, vistarin, iodio, eranol, 1L, CHEBI:17606, Brown, PNDPGZBMCMUPRI-UHFFFAOYSA-N, molecular iodine, 0 deg.C, 11355, 253.809, Amber glass bottle, Liquid, 807, II, 0.7, 14mmHg

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7553-56-2
Molecular Formula I2
Molecular Weight (g/mol) 253.81
MDL Number MFCD00011355 MFCD00164163
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
Synonym iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
PubChem CID 807
ChEBI CHEBI:17606
IUPAC Name molecular iodine
SMILES II

Specifications

Color Brown
Melting Point 0°C
Grade pH.Eur.
Packaging Amber glass bottle
Vapor Pressure 14mmHg
Vapor Density 0.7
pH 5
Quantity 1 L
Physical Form Liquid
compliance-icons
Product Identifier
  • Iodine solution 0.05M (0.1N)
Signal Word
  • Warning
Hazard Category
  • Specific target organ toxicity Category 2
Hazard Statement
  • H373-May cause damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P314-Get medical advice/attention if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Potassium iodide, Iodine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.