missing translation for 'onlineSavingsMsg'
Learn More

Iodine Solution 0.05M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™

Product Code. 10375800 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
Unit Size:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10375800 2.5 L 2.5 L
10625112 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10375800 Supplier Fisher Chemical Supplier No. J/4410/17
Discount Lab Supplies
Use promo code 28170
View all promotions
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

I2, CAS Number-7553-56-2, tincture iodine, iodine sublimed, iodine solution, iodine crystals, iodine, iode, diiodine, vistarin, iodio, eranol, 1L, CHEBI:17606, Brown, PNDPGZBMCMUPRI-UHFFFAOYSA-N, molecular iodine, 0 deg.C, 11355, 253.809, Amber glass bottle, Liquid, 807, II, 0.7, 14mmHg

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7553-56-2
Molecular Formula I2
Molecular Weight (g/mol) 253.81
MDL Number MFCD00011355 MFCD00164163
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
Synonym iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
PubChem CID 807
ChEBI CHEBI:17606
IUPAC Name molecular iodine
SMILES II

Specifications

Color Brown
Melting Point 0°C
Packaging Amber glass bottle
Vapor Pressure 14mmHg
Vapor Density 0.7
pH 5
Quantity 2.5 L
Physical Form Liquid
compliance-icons
Product Identifier
  • Iodine solution 0.05M (0.1N)
Signal Word
  • Warning
Hazard Category
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P314-Get medical advice/attention if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Iodine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.