missing translation for 'onlineSavingsMsg'
Learn More

Iodine, resublimed crystals, ACS, 99.8%

Product Code. p-4407455
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 11314947

Brand: Thermo Scientific Alfa Aesar 041955.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7553-56-2 | I2 | 253.81 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7553-56-2
Molecular Formula I2
Molecular Weight (g/mol) 253.81
MDL Number MFCD00011355 MFCD00164163
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
Synonym iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
PubChem CID 807
ChEBI CHEBI:17606
IUPAC Name molecular iodine
SMILES II

Specifications

Boiling Point 183°C to 186° (sublimation)
Color Gray-Purple
Physical Form Prills or Crystals
Quantity 100 g
Assay Percent Range 99.8%
Formula Weight 253.81
Grade ACS
Odor Irritating
Appearance Bluish-black
Density 4.930 g/cm3
Chemical Name or Material Iodine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Iodine, resublimed crystals
Signal Word
  • Warning
Hazard Category
  • ACUTE AQUATIC Acute 1
  • Acute toxicity Category 4
Hazard Statement
  • H312 + H332-Harmful in contact with skin or if inhaled.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.