missing translation for 'onlineSavingsMsg'
Learn More

Iodine, resublimed crystals, Puratronic™, 99.9985% (metals basis)

Product Code. 11380789
Click to view available options
Quantity:
2.5 g
25 g
100 g
Unit Size:
100g
2.5g
25g
This item is not returnable. View return policy

Product Code. 11380789

Brand: Thermo Scientific Alfa Aesar 010619.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7553-56-2 | I2 | 253.81 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7553-56-2
Molecular Formula I2
Molecular Weight (g/mol) 253.81
MDL Number MFCD00011355 MFCD00164163
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
Synonym iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
PubChem CID 807
ChEBI CHEBI:17606
SMILES II

Specifications

Boiling Point 183°C to 186°C
Melting Point 183°C to 186° (sublimation)
Color Black-Blue
Physical Form Solid
Quantity 25 g
Assay Percent Range 99.9985% (Metals basis)
UN Number 3495
IUPAC Name molecular iodine
Formula Weight 253.81
Odor Irritating
Appearance Bluish-black crystals
Density 4.93 g/cm3 at 20°C
Chemical Name or Material Iodine
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.