missing translation for 'onlineSavingsMsg'
Learn More

Iodine, Puriss., ≥99.5% (RT), particles (round), Solstice

Product Code. 15625870 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
250 g
1 kg
Packaging:
Glass Bottle
Unit Size:
100g
1kg
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15625870 250 g Glass Bottle 250g
15605870 100 g Glass Bottle 100g
15615870 1 kg Glass Bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15625870 Supplier Honeywell-Fluka Supplier No. 57655250G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Puriss.,≥99.5% (RT), particles (round)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 7553-56-2
Molecular Formula I2
Molecular Weight (g/mol) 253.81
MDL Number MFCD00011355 MFCD00164163
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
Synonym iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
PubChem CID 807
ChEBI CHEBI:17606
SMILES II

Spezifikation

Boiling Point 184°C (literature)
Melting Point 113°C (literature)
CAS Max % 100.0000%
Quantity 250 g
Linear Formula I2
UN Number UN3495
Beilstein 3587194
IUPAC Name molecular iodine
Formula Weight 253.81
Percent Purity ≥99.5%
Grade puriss.
Packaging Glass Bottle
Density 4.930 g/cm3
Chemical Name or Material Iodine, Puriss., ≥99.5% (RT), particles (round)
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.