missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodine monochloride, approx. 0.22N soln. in glacial acetic acid, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 mL
1 L
4 L
Description
For determination of iodine absorption number of fatsIodine monochloride is used as a catalyst in organic synthesis. It is the source of electrophilic iodine in the synthesis of certain aromatic iodides. It is used to determine the iodine value of a substance.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Moisture and Light Sensitive
Specifications
Physical Form | Liquid |
Quantity | 500 mL |
Solubility Information | Miscible with water,alcohol,ether and carbon disulfide. |
Formula Weight | 162.36 |
Ignition Point | 485°C (905°F) (acetic acid) |
Sensitivity | Light and moisture sensitive |
Appearance | Liquid |
Chemical Name or Material | Iodine monochloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'