missing translation for 'onlineSavingsMsg'
Learn More

Iodine monobromide, 98% min

Product Code. p-6827158
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11303488

Brand: Thermo Scientific Alfa Aesar 039106.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7789-33-5 | BrI | 206.81 g/mol

Iodine bromide is used for bromination of and for determining iodine used. IBr is used in some types of iodometry. It is also used as a organic synthesis reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-33-5
Molecular Formula BrI
Molecular Weight (g/mol) 206.81
MDL Number MFCD00011353
InChI Key CBEQRNSPHCCXSH-UHFFFAOYSA-N
Synonym iodine monobromide, iodine bromide, iodobromine, iodine bromide ibr, unii-g0k622rd9n, iodinemonobromide, iodobromane, bromoiodine, jodmonobromid, hanus solution
PubChem CID 82238
IUPAC Name iodobromane
SMILES BrI

Specifications

Boiling Point 116°C (decomposition)
Melting Point 50° (sublimation)
Physical Form Crystalline
Quantity 25 g
Assay Percent Range 98% min
Beilstein 3902973
Flash Point None
Solubility Information It is soluble in alcohol,water,ether,carbon disulfi,and glacial acetic acid.
Formula Weight 206.81
Sensitivity Light and moisture sensitive
Density 4.416
Chemical Name or Material Iodine bromide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Iodine monobromide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Iodine bromide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.