missing translation for 'onlineSavingsMsg'
Learn More

Iodine, Honeywell™

Product Code. 15695860 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15695860 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15695860 Supplier Honeywell-Fluka Supplier No. 57652100G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Tested according to Ph.Eur.

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

CAS 7553-56-2
Quantity 100 g
Molecular Formula I2
MDL Number MFCD00011355 MFCD00164163
UN Number UN3495
Beilstein 3587194
Synonym iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
SMILES II
IUPAC Name molecular iodine
Molecular Weight (g/mol) 253.81
PubChem CID 807
ChEBI CHEBI:17606
Formula Weight 253.81g/mol
Percent Purity 99%
Vapor Pressure 1 mmHg (38.7°C), 0.31 mmHg (25°C)
Density Vapor Density: 9 (vs air)
Chemical Name or Material Iodine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.