Learn More
Thermo Scientific Chemicals Iodine, 0.1N Standardized Solution
Description
Iodine, 0.1N standardized solution is used for accurate volumetric analysis. It is also used as a disinfectant and antibacterial agent in solutions, tinctures and salves.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Physical Form | Liquid |
| Solubility | Miscible with chloroform, carbon tetrachloride, carbon disulfide, cyclohexane, methanol, ethyl acetate, toluene, benzene, n-hexane, butan-2-ol, bromoethane, n-heptane, glycerol and diethyl ether |
| IUPAC Name | diiodine |
| Quantity | 4 L |
| Formula Weight | 253.81 |
| Concentration or Composition (by Analyte or Components) | Iodine: 1.3%, Potassium iodide: 2.5%, Water: 96.2% |
| Chemical Name or Material | Iodine |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.