missing translation for 'onlineSavingsMsg'
Learn More

Iodic Acid, TRC

Product Code. 30388936
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30388936 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30388936 Supplier Toronto Research Chemicals Supplier No. TRCI666725100G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Iodic Acid
CAS 7782-68-5
Quantity 100 g
Molecular Formula HIO3
Synonym Hydrogen Iodate, Hydrogen Iodate (HIO3)
InChI Formula InChI=1S/HIO3/c2-1(3)4/h(H,2,3,4)
SMILES OI(=O)=O
IUPAC Name iodic acid
Molecular Weight (g/mol) 175.91
Formula Weight 175.897
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.