missing translation for 'onlineSavingsMsg'
Learn More

Iodic acid, ACS, 99.5% min, Thermo Scientific Chemicals

Catalog Number 11355019
Click to view available options
Quantity:
50 g
250 g
7782-68-5
HIO3
175.909
MFCD00011348
ICIWUVCWSCSTAQ-UHFFFAOYSA-N
unii-6u8j18jsbm, 6u8j18jsbm, iodicacid, monoiodic acid, caswell no. 499a, iodsaeure, trioxoiodic acid, iodic v acid, sodium iodate min
24345
CHEBI:24857
iodic acid
OI(=O)=O
7782-68-5
HIO3
175.909
MFCD00011348
ICIWUVCWSCSTAQ-UHFFFAOYSA-N
unii-6u8j18jsbm, 6u8j18jsbm, iodicacid, monoiodic acid, caswell no. 499a, iodsaeure, trioxoiodic acid, iodic v acid, sodium iodate min
24345
CHEBI:24857
iodic acid
OI(=O)=O

Iodic acid is used in analytical chemistry laboratories to standardize solutions of both weak and strong bases using methyl red or methyl orange as the indicator. It acts as a key starting material to synthesize sodium or potassium iodate, thereby increasing the iodine content of salt.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Appearance Powder/chunks
Melting Point 110°C (decomposition)
Density 4.629 g/cm3
Sensitivity Light sensitive
Assay Percent Range 99.5%
Quantity 50 g
Physical Form Powder/Chunks
Solubility Information Soluble in water and methanol.
Formula Weight 175.91
Grade ACS
Chemical Name or Material Iodic Acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Iodic acid, ACS
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Iodic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'