missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Involucrin Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 28462-1-AP-20UL
This item is not returnable.
View return policy
Description
The Involucrin antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Involucrin. This antibody recognizes human antigen. The Involucrin antibody has been validated for the following applications: WB, IHC, IF/ICC, FC (Intra), ELISA analysis.Specifications
| Involucrin | |
| Polyclonal | |
| Unconjugated | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human | |
| 120 kDa | |
| Liquid |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Flow Cytometry (Intracellular Staining), ELISA | |
| 450 μg/mL | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 3713 | |
| Store at -20°C. Stable for one year after shipment. | |
| Polyclonal Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering