missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Involucrin Rabbit anti-Human, Mouse, Rat, Clone: 8H7B3, Novus Biologicals™
Description
Involucrin Monoclonal antibody specifically detects Involucrin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | Involucrin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | 8H7B3 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin |
| Formulation | PBS, 0.05% BSA, 50% glycerol, pH7.3 |
| Gene Alias | involucrin |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide corresponding to a sequence within amino acids 100-200 of human Involucrin (P07476). KAENPEQQLKQEKTQRDQQLNKQLEEEKKLLDQQLDQELVKRDEQLGMKKEQLLELPEQQEGHLKHLEQQEGQLKHPEQQEGQLELPEQQEGQLELPEQQE |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?