missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Involucrin Mouse anti-Human, Porcine, Canine, Primate, Monkey, Clone: rIVRN/827, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07559-100ug
This item is not returnable.
View return policy
Description
Involucrin Monoclonal specifically detects Involucrin in Human, Porcine, Canine, Primate, Monkey samples. It is validated for Immunohistochemistry-Paraffin.
Specifications
| Involucrin | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry-Paraffin 0.1-0.2 μg/mL | |
| involucrin | |
| Recombinant human full-length Involucrin protein (IVRN) | |
| 100 μg | |
| Extracellular Matrix | |
| 3713 | |
| Store at 4°C. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| rIVRN/827 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Pig, Canine, Primate, Monkey | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction