missing translation for 'onlineSavingsMsg'
Learn More

Involucrin Antibody (rIVRN/827) - Azide and BSA Free, Novus Biologicals™

Product Code. 18319101 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18319101 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18319101 Supplier Novus Biologicals Supplier No. NBP308292

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Involucrin Monoclonal specifically detects Involucrin in Human, Porcine, Canine, Primate, Monkey samples. It is validated for Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Involucrin
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone rIVRN/827
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin : 0.1-0.2 ug/mL
Formulation 10mM PBS
Gene Alias involucrin
Host Species Mouse
Immunogen Recombinant human full-length Involucrin protein (IVRN)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Extracellular Matrix
Primary or Secondary Primary
Gene ID (Entrez) 3713
Target Species Human, Pig, Canine, Primate, Monkey
Content And Storage Store at -20 to -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.