missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Zero Blunt™ TOPO™ PCR Cloning Kit, with One Shot™ MAX Efficiency DH5α-T1R E. coli

Product Code. 10525383
Click to view available options
Quantity:
25 Reactions
50 Reactions
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10525383

Brand: Invitrogen™ K282040

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

Linearized and topoisomerase I-activated pCR™Blunt II-TOPO™ vector; Salt solution; dNTPs; Control template; M13 forward and reverse primers; Sterile water; One Shot™ Chemically Competent or Electrocomp™ E. coli; S.O.C. medium; Supercoiled pUC19 control plasmid

Offers the fastest and easiest method for high-efficiency (cloning of blunt-end PCR products amplified with proofreading thermostable polymerases

Kit includes linearized and topoisomerase I-activated pCR™-Blunt II-TOPO Vector for 5-minute benchtop ligations without ligase.

Selected Zero Blunt TOPO PCR Cloning Kits are available in combo kits combined with a PureLink™ Quick Plasmid Miniprep Kit (50 preps) for fast plasmid purification of your TOPO-cloned inserts for downstream analysis.

  • ccdB gene for positive selection
  • EcoR I sites flanking the PCR product insertion site for easy excision of inserts
  • Kanamycin and Zeocin™ resistance genes for your choice of selection in E. coli
  • SP6 promoter/primer site for in vitro RNA transcription and sequencing
  • M13 forward and reverse primer sites for sequencing or PCR screening

Applications:

Cloning, PCR Cloning

TRUSTED_SUSTAINABILITY

Specifications

Bacterial or Yeast Strain DH5α
Cloning Method Blunt TOPO™
Content And Storage Box 1:
• Linearized and topoisomerase I-activated pCR™Blunt II-TOPO™ vector
• Salt solution
• dNTPs
• Control template
• M13 forward and reverse primers
• Sterile water

Store at -5 to -30°C.
All reagents are stable for 6 months when properly stored.

Box 2:
• One Shot™ Chemically Competent or Electrocomp™ E. coli
• S.O.C. medium
• Supercoiled pUC19 control plasmid

Store at -68 to -85°C.
Format Kit
Product Line One Shot
Product Type PCR Cloning Kit
Promoter SP6
Quantity 50 Reactions
Vector Blunt DNA Cloning Vectors

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.