missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UltraPure™ Phenol:Chloroform:Isoamyl Alcohol (25:24:1, v/v)
Description
- Reagent consists of highly pure chloroform, isoamyl alcohol, and UltraPure™ Phenol saturated with Tris-HCl
- When mixtures are extracted with Phenol:Chloroform:Isoamyl Alcohol, proteins are denatured and collected in the organic phase or at the interphase, while nucleic acids remain in the aqueous phase
- UltraPure™ Phenol:Chloroform:lsoamyl Alcohol contains no preservatives
- It is packaged under an inert gas in shatter-resistant, plastic-coated amber bottles
- The appearance of the solution is evaluated at room temperature
- No RNase or DNase activity is detected
- Store in the refrigerator at 2°C to 8°C
Order Info
Shipping Condition: Room temperature
Specifications
Specifications
| Chemical Name or Material | Phenols |
| Content And Storage | Store in refrigerator (2–8°C). |
| pH | 8.05 |
| Product Line | UltraPure |
| Purity | Molecular Biology Grade |
| Quantity | 400 mL |
| Shipping Condition | Room Temperature |
| Form | Liquid |
Safety and Handling
Product Identifier
- UltraPure Phenol:Chloroform:IsoamylAlcohol(25:24:1,v/v)
- Danger
- Acute toxicity Category 1
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Germ cell mutagenicity Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 1
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
- H341-Suspected of causing genetic defects.
- H351-Suspected of causing cancer.
- H361-Suspected of damaging fertility or the unborn child.
- H372-Causes damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
- P201-Obtain special instructions before use.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash hands thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contain: Chloroform, Phenol,3-methyl-1-butanol
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?