missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UltraPure™ N,N'-Methylenebisacrylamide
Description
- UltraPure N,N′-Methylenebisacrylamide is most frequently used cross-linking agent for polyacrylamide gels
Acrylamide Gel Electrophoresis, DNA & RNA Purification & Analysis, Gel Casting, Nucleic Acid Gel Electrophoresis & Blotting, Protein Gel Electrophoresis, Proteins, Expression, Isolation & Analysis
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Content And Storage | Store at room temperature. |
| For Use With (Application) | Gel Electrophoresis |
| Product Line | UltraPure |
| Product Type | N,N'-Methylenebisacrylamide |
| Quantity | 100 g |
| Shipping Condition | Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?