missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UltraPure™ Glycine

Product Code. 11568746
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11568746 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 11568746 Supplier Invitrogen™ Supplier No. 15527013

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Suitable for buffering at extremes of common buffering range

  • UltraPure Glycine is amino acid, with pKa1 of 2.3 and a pKa2 of 9.6, used in preparation of some electrophoresis buffers

1D Gel Electrophoresis, ChIP-on-Chip, Chromatin Biology, Chromatin Immunoprecipitation (ChIP), Protein Gel Electrophoresis, Proteins, Expression, Isolation & Analysis, RNAi, Epigenetics & Non-Coding RNA Research

Order Info

Shipping Condition: Approved for shipment at room temperature or on wet ice

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Glycine
Content And Storage Store at 15°C to 30°C.
Product Line UltraPure
Product Type Glycine
Quantity 500 g
Purity Molecular Biology Grade
Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
Form Powder

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.